missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-2-phenylacetophenone, 97%

Catalog No. AC403170250
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC403170250 25 g Glass bottle
AC403170050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC403170250 Supplier Thermo Scientific Chemicals Supplier No. 403170250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1484-50-0 | C14H11BrO | 275.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1484-50-0
Molecular Formula C14H11BrO
Molecular Weight (g/mol) 275.13
MDL Number MFCD00000136
InChI Key ZFFBIQMNKOJDJE-UHFFFAOYSA-N
PubChem CID 102630
IUPAC Name 2-bromo-1,2-diphenylethanone
SMILES C1=CC=C(C=C1)C(C(=O)C2=CC=CC=C2)Br

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 53°C to 58°C
Color Beige to Brown
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 25 g
Synonym 2-bromo-2-phenylacetophenone, desyl bromide, 2-bromo-1,2-diphenylethan-1-one, ethanone, 2-bromo-1,2-diphenyl, ethanone,2-bromo-1,2-diphenyl, 2-bromo-1,2-diphenyl-ethanone, desylbromide, alpha-bromodeoxybenzoin, alpha-bromodesoxibenzoine
Formula Weight 275.13
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromo-2-phenylacetophenone, 97%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
EINECSNumber 216-057-1
TSCA TSCA

RUO – Research Use Only