Learn More
2-Bromo-2'-nitroacetophenone, 98%, Thermo Scientific™
$330.24
Chemical Identifiers
| CAS | 6851-99-6 |
|---|---|
| Molecular Formula | C8H6BrNO3 |
| Molecular Weight (g/mol) | 244.4 |
| MDL Number | MFCD00010294 |
| InChI Key | SGXUUCSRVVSMGK-UHFFFAOYSA-N |
| Synonym | 2-bromo-2'-nitroacetophenone, 2'-nitrophenacyl bromide, 2-bromo-1-2-nitrophenyl ethanone, 2-nitrophenacyl bromide, 2-bromo-1-2-nitro-phenyl-ethanone, o-nitrophenacyl bromide, o-nitrophenethyl bromide, 2-o-nitrophenyl ethyl bromide, 2-bromo-1-2-nitrophenyl ethan-1-one, ethanone, 2-bromo-1-2-nitrophenyl |
| PubChem CID | 244025 |
| IUPAC Name | 2-bromo-1-(2-nitrophenyl)ethanone |
| SMILES | C1=CC=C(C(=C1)C(=O)CBr)[N+](=O)[O-] |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
AC398850050
|
N/A | 5g | Glass bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| C8H6BrNO3 | |
| MFCD00010294 | |
| 2-bromo-2'-nitroacetophenone, 2'-nitrophenacyl bromide, 2-bromo-1-2-nitrophenyl ethanone, 2-nitrophenacyl bromide, 2-bromo-1-2-nitro-phenyl-ethanone, o-nitrophenacyl bromide, o-nitrophenethyl bromide, 2-o-nitrophenyl ethyl bromide, 2-bromo-1-2-nitrophenyl ethan-1-one, ethanone, 2-bromo-1-2-nitrophenyl | |
| 2-bromo-1-(2-nitrophenyl)ethanone |
Specifications
| 6851-99-6 | |
| 100.0 | |
| 97.5% min. (GC) | |
| O2NC6H4COCH2Br | |
| 2-bromo-2'-nitroacetophenone, 2'-nitrophenacyl bromide, 2-bromo-1-2-nitrophenyl ethanone, 2-nitrophenacyl bromide, 2-bromo-1-2-nitro-phenyl-ethanone, o-nitrophenacyl bromide, o-nitrophenethyl bromide, 2-o-nitrophenyl ethyl bromide, 2-bromo-1-2-nitrophenyl ethan-1-one, ethanone, 2-bromo-1-2-nitrophenyl | |
| C1=CC=C(C(=C1)C(=O)CBr)[N+](=O)[O-] | |
| 244.4 | |
| 244.4 | |
| 2-Bromo-2'-nitroacetophenone |
| 97.5 | |
| Authentic | |
| C8H6BrNO3 | |
| MFCD00010294 | |
| SGXUUCSRVVSMGK-UHFFFAOYSA-N | |
| 2-bromo-1-(2-nitrophenyl)ethanone | |
| 244025 | |
| 98% |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF ON SKIN (
GHS Signal Word: Danger
RUO – Research Use Only