missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-2'-nitroacetophenone, 98%

Catalog No. AAH3027503
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3027503 1 g
AAH3027506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3027503 Supplier Thermo Scientific Chemicals Supplier No. H3027503
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 6851-99-6 | C8H6BrNO3 | 244.044 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6851-99-6
Molecular Formula C8H6BrNO3
Molecular Weight (g/mol) 244.044
MDL Number MFCD00010294
InChI Key SGXUUCSRVVSMGK-UHFFFAOYSA-N
Synonym 2-bromo-2'-nitroacetophenone, 2'-nitrophenacyl bromide, 2-bromo-1-2-nitrophenyl ethanone, 2-nitrophenacyl bromide, 2-bromo-1-2-nitro-phenyl-ethanone, o-nitrophenacyl bromide, o-nitrophenethyl bromide, 2-o-nitrophenyl ethyl bromide, 2-bromo-1-2-nitrophenyl ethan-1-one, ethanone, 2-bromo-1-2-nitrophenyl
PubChem CID 244025
IUPAC Name 2-bromo-1-(2-nitrophenyl)ethanone
SMILES C1=CC=C(C(=C1)C(=O)CBr)[N+](=O)[O-]

Specifications

Melting Point 53°C to 56°C
Odor Pungent
Quantity 1 g
UN Number UN3261
Formula Weight 244.05
Percent Purity 98%
Chemical Name or Material 2-Bromo-2'-nitroacetophenone
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only