missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-2-nitro-1,3-propanediol 98.0+%, TCI America™

Catalog No. B124725G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B124725G 25 g
B1247100G 100 g
B1247500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B124725G Supplier TCI America Supplier No. B124725G

Chemical Identifiers

CAS 52-51-7
Molecular Formula C3H6BrNO4
Molecular Weight (g/mol) 199.988
MDL Number MFCD00007390
InChI Key LVDKZNITIUWNER-UHFFFAOYSA-N
Synonym bronopol, 2-bromo-2-nitro-1,3-propanediol, bronosol, bronocot, bronidiol, bronopolu, bronotak, 1,3-propanediol, 2-bromo-2-nitro, onyxide 500, lexgard bronopol
PubChem CID 2450
ChEBI CHEBI:31306
IUPAC Name 2-bromo-2-nitropropane-1,3-diol
SMILES C(C(CO)([N+](=O)[O-])Br)O

Specifications

Melting Point 131°C
Quantity 25 g
UN Number 3241
Formula Weight 199.99
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromo-2-nitro-1,3-propanediol
EINECSNumber (2)-0325
RTECSNumber TY3385000
TSCA Yes