missing translation for 'onlineSavingsMsg'
Learn More

Sigma Aldrich 2-Bromo-2-nitro-1,3-propanediol

Catalog No. 134708100G
Encompass
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
134708100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 134708100G Supplier Sigma Aldrich Supplier No. 134708100G
Only null left

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

2-Bromo-2-nitro-1,3-propanediol

Specifications

CAS 52-51-7
Melting Point 130°C to 133°C (lit.)
Molecular Formula C3H6NO4Br
Linear Formula HOCH2C(Br)(NO2)CH2OH
MDL Number MFCD00007390
Quantity 100 g
Synonym BNPD; BNPK; Bronopol
Molecular Weight (g/mol) 199.99
Percent Purity 98%