missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromo-2'-methylacetophenone 96.0+%, TCI America™
$48.46 - $158.44
Chemical Identifiers
| CAS | 51012-65-8 |
|---|---|
| Molecular Formula | C9H9BrO |
| Molecular Weight (g/mol) | 213.074 |
| MDL Number | MFCD04038955 |
| InChI Key | XMGAXELQRATLJP-UHFFFAOYSA-N |
| Synonym | 2-bromo-1-o-tolyl ethanone, 2-bromo-1-2-methylphenyl ethan-1-one, 2'-methylphenacyl bromide, 2-bromo-1-o-tolylethanone, 2-bromo-1-2-methylphenyl ethanone, 2-methylphenacyl bromide, 2-bromo-1-o-tolyl-ethanone, 2-bromo-2'-methylacetophenone, ethanone, 2-bromo-1-methylphenyl, 2-bromoacetyl toluene |
| PubChem CID | 12430676 |
| IUPAC Name | 2-bromo-1-(2-methylphenyl)ethanone |
| SMILES | CC1=CC=CC=C1C(=O)CBr |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
B4750200MG
|
TCI America
B4750200MG |
200 mg |
Each for $48.46
|
|
|||||
|
B47501G
|
TCI America
B47501G |
1 g |
Each for $158.44
|
|
|||||
Chemical Identifiers
| 51012-65-8 | |
| 213.074 | |
| XMGAXELQRATLJP-UHFFFAOYSA-N | |
| 12430676 | |
| CC1=CC=CC=C1C(=O)CBr |
| C9H9BrO | |
| MFCD04038955 | |
| 2-bromo-1-o-tolyl ethanone, 2-bromo-1-2-methylphenyl ethan-1-one, 2'-methylphenacyl bromide, 2-bromo-1-o-tolylethanone, 2-bromo-1-2-methylphenyl ethanone, 2-methylphenacyl bromide, 2-bromo-1-o-tolyl-ethanone, 2-bromo-2'-methylacetophenone, ethanone, 2-bromo-1-methylphenyl, 2-bromoacetyl toluene | |
| 2-bromo-1-(2-methylphenyl)ethanone |
Specifications
| 51012-65-8 | |
| 83°C | |
| MFCD04038955 | |
| 2810 | |
| XMGAXELQRATLJP-UHFFFAOYSA-N | |
| 2-bromo-1-(2-methylphenyl)ethanone | |
| 12430676 | |
| ≥96.0% (GC,T) | |
| 2-Bromo-2′-methylacetophenone |
| Yellow | |
| C9H9BrO | |
| 200 mg | |
| 2-bromo-1-o-tolyl ethanone, 2-bromo-1-2-methylphenyl ethan-1-one, 2'-methylphenacyl bromide, 2-bromo-1-o-tolylethanone, 2-bromo-1-2-methylphenyl ethanone, 2-methylphenacyl bromide, 2-bromo-1-o-tolyl-ethanone, 2-bromo-2'-methylacetophenone, ethanone, 2-bromo-1-methylphenyl, 2-bromoacetyl toluene | |
| CC1=CC=CC=C1C(=O)CBr | |
| 213.074 | |
| 213.07 | |
| Liquid |
Safety and Handling
TSCA : No