missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromo-2'-methylacetophenone 96.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Boiling Point | 83°C |
| Quantity | 200 mg |
| UN Number | 2810 |
| Formula Weight | 213.07 |
| Percent Purity | ≥96.0% (GC,T) |
| Physical Form | Liquid |
| Chemical Name or Material | 2-Bromo-2′-methylacetophenone |
Safety and Handling
| TSCA | No |