missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-2'-methylacetophenone, 98%

Catalog No. AAH6465106
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6465106 5 g
AAH6465103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6465106 Supplier Thermo Scientific Chemicals Supplier No. H6465106
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 51012-65-8 | C9H9BrO | 213.074 g/mol

2-Bromo-2'-methylacetophenone is an aromatic building block for proteomics research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromo-2′-methylacetophenone is an aromatic building block for proteomics research.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.

Chemical Identifiers

CAS 51012-65-8
Molecular Formula C9H9BrO
Molecular Weight (g/mol) 213.074
MDL Number MFCD04038955
InChI Key XMGAXELQRATLJP-UHFFFAOYSA-N
Synonym 2-bromo-1-o-tolyl ethanone, 2-bromo-1-2-methylphenyl ethan-1-one, 2'-methylphenacyl bromide, 2-bromo-1-o-tolylethanone, 2-bromo-1-2-methylphenyl ethanone, 2-methylphenacyl bromide, 2-bromo-1-o-tolyl-ethanone, 2-bromo-2'-methylacetophenone, ethanone, 2-bromo-1-methylphenyl, 2-bromoacetyl toluene
PubChem CID 12430676
IUPAC Name 2-bromo-1-(2-methylphenyl)ethanone
SMILES CC1=CC=CC=C1C(=O)CBr

Specifications

Boiling Point 81°C to 83°C
Quantity 5 g
Beilstein 508116
Solubility Information Slightly soluble in water.
Formula Weight 213.07
Percent Purity 98%
Chemical Name or Material 2-Bromo-2'-methylacetophenone
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only