missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromo-2'-methylacetophenone, 98%
Description
2-Bromo-2'-methylacetophenone is an aromatic building block for proteomics research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Bromo-2′-methylacetophenone is an aromatic building block for proteomics research.
Solubility
Slightly soluble in water.
Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
2-Bromo-2′-methylacetophenone is an aromatic building block for proteomics research.
Solubility
Slightly soluble in water.
Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Specifications
Specifications
| Boiling Point | 81°C to 83°C |
| Quantity | 1 g |
| Beilstein | 508116 |
| Solubility Information | Slightly soluble in water. |
| Formula Weight | 213.07 |
| Percent Purity | 98% |
| Chemical Name or Material | 2-Bromo-2'-methylacetophenone |
Safety and Handling
| Hazard Category | H315-H319-H335 |
| Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only