missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-2'-hydroxyacetophenone, 97%

Catalog No. p-7028415
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2572206 5 g
AAB2572214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2572206 Supplier Thermo Scientific Chemicals Supplier No. B2572206
Only null left

CAS: 2491-36-3 | C8H7BrO2 | 215.046 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2491-36-3
Molecular Formula C8H7BrO2
Molecular Weight (g/mol) 215.046
MDL Number MFCD01727570
InChI Key SGPKEYSZPHMVNI-UHFFFAOYSA-N
PubChem CID 200671
IUPAC Name 2-bromo-1-(2-hydroxyphenyl)ethanone
SMILES C1=CC=C(C(=C1)C(=O)CBr)O

Specifications

Melting Point 45°C to 47°C
Quantity 5 g
UN Number UN3263
Synonym 2-bromo-2'-hydroxyacetophenone, 2-bromo-1-2-hydroxyphenyl ethanone, 2-hydroxyphenacyl bromide, ethanone, 2-bromo-1-2-hydroxyphenyl, 2-bromo-1-2-hydroxyphenyl ethan-1-one, acetophenone, 2-bromo-2'-hydroxy-6ci,7ci,8ci, ethanone,2-bromo-1-2-hydroxyphenyl, 2-bromoacetyl phenol, acmc-1cg90, 4-08-00-00329 beilstein handbook reference
Formula Weight 215.05
Percent Purity 97%
Chemical Name or Material 2-Bromo-2'-hydroxyacetophenone
Hazard Category H302-H314-H335
Hazard Statement GHS H Statement
H314-H318-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, BASIC, ORGANIC, N.O.S.
RTECSNumber KM5556400
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only