missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromo-2'-fluoroacetophenone 98.0+%, TCI America™
$257.97 - $1033.82
Chemical Identifiers
| CAS | 655-15-2 |
|---|---|
| Molecular Formula | C8H6BrFO |
| Molecular Weight (g/mol) | 217.037 |
| MDL Number | MFCD00278796 |
| InChI Key | QDNWNJSLWKHNTM-UHFFFAOYSA-N |
| Synonym | 2-bromo-2'-fluoroacetophenone, 2-bromo-1-2-fluorophenyl ethanone, 2-fluorophenacyl bromide, 2-bromo-1-2-fluorophenyl ethan-1-one, alpha-bromo-2-fluoroacetophenone, 2-fluorophenacylbromide, ethanone, 2-bromo-1-2-fluorophenyl, o-fluorophenacyl bromide, omega-bromo-2-fluoroacetophenone, 2-bromo-1-2'-fluorophenyl ethanone |
| PubChem CID | 2737449 |
| IUPAC Name | 2-bromo-1-(2-fluorophenyl)ethanone |
| SMILES | C1=CC=C(C(=C1)C(=O)CBr)F |
Chemical Identifiers
| 655-15-2 | |
| 217.037 | |
| QDNWNJSLWKHNTM-UHFFFAOYSA-N | |
| 2737449 | |
| C1=CC=C(C(=C1)C(=O)CBr)F |
| C8H6BrFO | |
| MFCD00278796 | |
| 2-bromo-2'-fluoroacetophenone, 2-bromo-1-2-fluorophenyl ethanone, 2-fluorophenacyl bromide, 2-bromo-1-2-fluorophenyl ethan-1-one, alpha-bromo-2-fluoroacetophenone, 2-fluorophenacylbromide, ethanone, 2-bromo-1-2-fluorophenyl, o-fluorophenacyl bromide, omega-bromo-2-fluoroacetophenone, 2-bromo-1-2'-fluorophenyl ethanone | |
| 2-bromo-1-(2-fluorophenyl)ethanone |
Specifications
| 655-15-2 | |
| 63°C | |
| MFCD00278796 | |
| 3261 | |
| QDNWNJSLWKHNTM-UHFFFAOYSA-N | |
| 2-bromo-1-(2-fluorophenyl)ethanone | |
| 2737449 | |
| ≥98.0% (GC) | |
| 2-Bromo-2′-fluoroacetophenone |
| White-Yellow | |
| C8H6BrFO | |
| 5 g | |
| 2-bromo-2'-fluoroacetophenone, 2-bromo-1-2-fluorophenyl ethanone, 2-fluorophenacyl bromide, 2-bromo-1-2-fluorophenyl ethan-1-one, alpha-bromo-2-fluoroacetophenone, 2-fluorophenacylbromide, ethanone, 2-bromo-1-2-fluorophenyl, o-fluorophenacyl bromide, omega-bromo-2-fluoroacetophenone, 2-bromo-1-2'-fluorophenyl ethanone | |
| C1=CC=C(C(=C1)C(=O)CBr)F | |
| 217.037 | |
| 217.04 | |
| Crystalline Lumps |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator