missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-2'-fluoroacetophenone 98.0+%, TCI America™

Catalog No. B51045G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B51045G 5 g
B510425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B51045G Supplier TCI America Supplier No. B51045G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 655-15-2
Molecular Formula C8H6BrFO
Molecular Weight (g/mol) 217.037
MDL Number MFCD00278796
InChI Key QDNWNJSLWKHNTM-UHFFFAOYSA-N
Synonym 2-bromo-2'-fluoroacetophenone, 2-bromo-1-2-fluorophenyl ethanone, 2-fluorophenacyl bromide, 2-bromo-1-2-fluorophenyl ethan-1-one, alpha-bromo-2-fluoroacetophenone, 2-fluorophenacylbromide, ethanone, 2-bromo-1-2-fluorophenyl, o-fluorophenacyl bromide, omega-bromo-2-fluoroacetophenone, 2-bromo-1-2'-fluorophenyl ethanone
PubChem CID 2737449
IUPAC Name 2-bromo-1-(2-fluorophenyl)ethanone
SMILES C1=CC=C(C(=C1)C(=O)CBr)F

Specifications

Color White-Yellow
Boiling Point 63°C
Quantity 5 g
UN Number 3261
Formula Weight 217.04
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 2-Bromo-2′-fluoroacetophenone
TSCA No
Recommended Storage Refrigerator