Learn More
2-Bromo-2'-fluoroacetophenone, 98%
CAS: 655-15-2 | C8H6BrFO | 217.037 g/mol
$67.79 - $393.66
Chemical Identifiers
| CAS | 655-15-2 |
|---|---|
| Molecular Formula | C8H6BrFO |
| Molecular Weight (g/mol) | 217.037 |
| MDL Number | MFCD00278796 |
| InChI Key | QDNWNJSLWKHNTM-UHFFFAOYSA-N |
| Synonym | 2-bromo-2'-fluoroacetophenone, 2-bromo-1-2-fluorophenyl ethanone, 2-fluorophenacyl bromide, 2-bromo-1-2-fluorophenyl ethan-1-one, alpha-bromo-2-fluoroacetophenone, 2-fluorophenacylbromide, ethanone, 2-bromo-1-2-fluorophenyl, o-fluorophenacyl bromide, omega-bromo-2-fluoroacetophenone, 2-bromo-1-2'-fluorophenyl ethanone |
| PubChem CID | 2737449 |
| IUPAC Name | 2-bromo-1-(2-fluorophenyl)ethanone |
| SMILES | C1=CC=C(C(=C1)C(=O)CBr)F |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH3417903
|
Thermo Scientific Chemicals
H3417903 |
1 g |
Each for $67.79
|
|
|||||
|
AAH3417909
|
Thermo Scientific Chemicals
H3417909 |
10 g |
Each for $393.66
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 655-15-2 | |
| 217.037 | |
| QDNWNJSLWKHNTM-UHFFFAOYSA-N | |
| 2737449 | |
| C1=CC=C(C(=C1)C(=O)CBr)F |
| C8H6BrFO | |
| MFCD00278796 | |
| 2-bromo-2'-fluoroacetophenone, 2-bromo-1-2-fluorophenyl ethanone, 2-fluorophenacyl bromide, 2-bromo-1-2-fluorophenyl ethan-1-one, alpha-bromo-2-fluoroacetophenone, 2-fluorophenacylbromide, ethanone, 2-bromo-1-2-fluorophenyl, o-fluorophenacyl bromide, omega-bromo-2-fluoroacetophenone, 2-bromo-1-2'-fluorophenyl ethanone | |
| 2-bromo-1-(2-fluorophenyl)ethanone |
Specifications
| 655-15-2 | |
| MFCD00278796 | |
| UN3265 | |
| QDNWNJSLWKHNTM-UHFFFAOYSA-N | |
| 2-bromo-1-(2-fluorophenyl)ethanone | |
| 2737449 | |
| 98% |
| C8H6BrFO | |
| 1 g | |
| 2-bromo-2'-fluoroacetophenone, 2-bromo-1-2-fluorophenyl ethanone, 2-fluorophenacyl bromide, 2-bromo-1-2-fluorophenyl ethan-1-one, alpha-bromo-2-fluoroacetophenone, 2-fluorophenacylbromide, ethanone, 2-bromo-1-2-fluorophenyl, o-fluorophenacyl bromide, omega-bromo-2-fluoroacetophenone, 2-bromo-1-2'-fluorophenyl ethanone | |
| C1=CC=C(C(=C1)C(=O)CBr)F | |
| 217.037 | |
| 217.04 | |
| 2-Bromo-2'-fluoroacetophenone |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H302+H312+H332-H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only