missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-2'-fluoroacetophenone, 98%

Catalog No. AAH3417909
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3417909 10 g
AAH3417903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3417909 Supplier Thermo Scientific Chemicals Supplier No. H3417909
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 655-15-2 | C8H6BrFO | 217.037 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 655-15-2
Molecular Formula C8H6BrFO
Molecular Weight (g/mol) 217.037
MDL Number MFCD00278796
InChI Key QDNWNJSLWKHNTM-UHFFFAOYSA-N
Synonym 2-bromo-2'-fluoroacetophenone, 2-bromo-1-2-fluorophenyl ethanone, 2-fluorophenacyl bromide, 2-bromo-1-2-fluorophenyl ethan-1-one, alpha-bromo-2-fluoroacetophenone, 2-fluorophenacylbromide, ethanone, 2-bromo-1-2-fluorophenyl, o-fluorophenacyl bromide, omega-bromo-2-fluoroacetophenone, 2-bromo-1-2'-fluorophenyl ethanone
PubChem CID 2737449
IUPAC Name 2-bromo-1-(2-fluorophenyl)ethanone
SMILES C1=CC=C(C(=C1)C(=O)CBr)F

Specifications

Quantity 10 g
UN Number UN3265
Formula Weight 217.04
Percent Purity 98%
Chemical Name or Material 2-Bromo-2'-fluoroacetophenone
Hazard Category H302+H312+H332-H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only