missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromo-2',4'-dimethoxyacetophenone 97.0+%, TCI America™
$411.98 - $1,259.07
Chemical Identifiers
| CAS | 60965-26-6 |
|---|---|
| Molecular Formula | C10H11BrO3 |
| Molecular Weight (g/mol) | 259.099 |
| MDL Number | MFCD00000197 |
| InChI Key | PKVBZABQCCQHLD-UHFFFAOYSA-N |
| Synonym | 2-bromo-1-2,4-dimethoxyphenyl ethanone, 2-bromo-2',4'-dimethoxyacetophenone, 2,4-dimethoxyphenacyl bromide, 2-bromo-1-2',4'-dimethoxyphenyl ethanone, 2-bromo-1-2,4-dimethoxyphenyl ethan-1-one, ethanone, 2-bromo-1-2,4-dimethoxyphenyl, 2-bromo-2',4'-dimethoxyacetopheneone, 1-2,4-dimethoxyphenyl-2-bromoethan-1-one, pubchem13433, acmc-1ba4l |
| PubChem CID | 98683 |
| IUPAC Name | 2-bromo-1-(2,4-dimethoxyphenyl)ethanone |
| SMILES | COC1=CC(=C(C=C1)C(=O)CBr)OC |
Chemical Identifiers
| 60965-26-6 | |
| 259.099 | |
| PKVBZABQCCQHLD-UHFFFAOYSA-N | |
| 98683 | |
| COC1=CC(=C(C=C1)C(=O)CBr)OC |
| C10H11BrO3 | |
| MFCD00000197 | |
| 2-bromo-1-2,4-dimethoxyphenyl ethanone, 2-bromo-2',4'-dimethoxyacetophenone, 2,4-dimethoxyphenacyl bromide, 2-bromo-1-2',4'-dimethoxyphenyl ethanone, 2-bromo-1-2,4-dimethoxyphenyl ethan-1-one, ethanone, 2-bromo-1-2,4-dimethoxyphenyl, 2-bromo-2',4'-dimethoxyacetopheneone, 1-2,4-dimethoxyphenyl-2-bromoethan-1-one, pubchem13433, acmc-1ba4l | |
| 2-bromo-1-(2,4-dimethoxyphenyl)ethanone |
Specifications
| 60965-26-6 | |
| C10H11BrO3 | |
| 1 g | |
| 2-bromo-1-2,4-dimethoxyphenyl ethanone, 2-bromo-2',4'-dimethoxyacetophenone, 2,4-dimethoxyphenacyl bromide, 2-bromo-1-2',4'-dimethoxyphenyl ethanone, 2-bromo-1-2,4-dimethoxyphenyl ethan-1-one, ethanone, 2-bromo-1-2,4-dimethoxyphenyl, 2-bromo-2',4'-dimethoxyacetopheneone, 1-2,4-dimethoxyphenyl-2-bromoethan-1-one, pubchem13433, acmc-1ba4l | |
| COC1=CC(=C(C=C1)C(=O)CBr)OC | |
| 259.099 | |
| 259.10 | |
| Crystalline Powder |
| 105°C | |
| MFCD00000197 | |
| 3261 | |
| PKVBZABQCCQHLD-UHFFFAOYSA-N | |
| 2-bromo-1-(2,4-dimethoxyphenyl)ethanone | |
| 98683 | |
| ≥97.0% (GC) | |
| 2-Bromo-2′,4′-dimethoxyacetophenone |
Safety and Handling
TSCA : No