missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-2',4'-dimethoxyacetophenone 97.0+%, TCI America™

Catalog No. B47681G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B47681G 1 g
B47685G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B47681G Supplier TCI America Supplier No. B47681G

Chemical Identifiers

CAS 60965-26-6
Molecular Formula C10H11BrO3
Molecular Weight (g/mol) 259.099
MDL Number MFCD00000197
InChI Key PKVBZABQCCQHLD-UHFFFAOYSA-N
Synonym 2-bromo-1-2,4-dimethoxyphenyl ethanone, 2-bromo-2',4'-dimethoxyacetophenone, 2,4-dimethoxyphenacyl bromide, 2-bromo-1-2',4'-dimethoxyphenyl ethanone, 2-bromo-1-2,4-dimethoxyphenyl ethan-1-one, ethanone, 2-bromo-1-2,4-dimethoxyphenyl, 2-bromo-2',4'-dimethoxyacetopheneone, 1-2,4-dimethoxyphenyl-2-bromoethan-1-one, pubchem13433, acmc-1ba4l
PubChem CID 98683
IUPAC Name 2-bromo-1-(2,4-dimethoxyphenyl)ethanone
SMILES COC1=CC(=C(C=C1)C(=O)CBr)OC

Specifications

Melting Point 105°C
Quantity 1 g
UN Number 3261
Formula Weight 259.10
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromo-2′,4′-dimethoxyacetophenone