Learn More
2-Bromo-2',4'-dimethoxyacetophenone, 98%
CAS: 60965-26-6 | C10H11BrO3 | 259.099 g/mol
$168.75 - $529.59
Chemical Identifiers
| CAS | 60965-26-6 |
|---|---|
| Molecular Formula | C10H11BrO3 |
| Molecular Weight (g/mol) | 259.099 |
| MDL Number | MFCD00000197 |
| InChI Key | PKVBZABQCCQHLD-UHFFFAOYSA-N |
| Synonym | 2-bromo-1-2,4-dimethoxyphenyl ethanone, 2-bromo-2',4'-dimethoxyacetophenone, 2,4-dimethoxyphenacyl bromide, 2-bromo-1-2',4'-dimethoxyphenyl ethanone, 2-bromo-1-2,4-dimethoxyphenyl ethan-1-one, ethanone, 2-bromo-1-2,4-dimethoxyphenyl, 2-bromo-2',4'-dimethoxyacetopheneone, 1-2,4-dimethoxyphenyl-2-bromoethan-1-one, pubchem13433, acmc-1ba4l |
| PubChem CID | 98683 |
| IUPAC Name | 2-bromo-1-(2,4-dimethoxyphenyl)ethanone |
| SMILES | COC1=CC(=C(C=C1)C(=O)CBr)OC |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAB2230003
|
Thermo Scientific Chemicals
B2230003 |
1 g |
Each for $168.75
|
|
|||||
|
AAB2230006
|
Thermo Scientific Chemicals
B2230006 |
5 g |
Each for $529.59
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 60965-26-6 | |
| 259.099 | |
| PKVBZABQCCQHLD-UHFFFAOYSA-N | |
| 98683 | |
| COC1=CC(=C(C=C1)C(=O)CBr)OC |
| C10H11BrO3 | |
| MFCD00000197 | |
| 2-bromo-1-2,4-dimethoxyphenyl ethanone, 2-bromo-2',4'-dimethoxyacetophenone, 2,4-dimethoxyphenacyl bromide, 2-bromo-1-2',4'-dimethoxyphenyl ethanone, 2-bromo-1-2,4-dimethoxyphenyl ethan-1-one, ethanone, 2-bromo-1-2,4-dimethoxyphenyl, 2-bromo-2',4'-dimethoxyacetopheneone, 1-2,4-dimethoxyphenyl-2-bromoethan-1-one, pubchem13433, acmc-1ba4l | |
| 2-bromo-1-(2,4-dimethoxyphenyl)ethanone |
Specifications
| 60965-26-6 | |
| C10H11BrO3 | |
| 1 g | |
| 2-bromo-1-2,4-dimethoxyphenyl ethanone, 2-bromo-2',4'-dimethoxyacetophenone, 2,4-dimethoxyphenacyl bromide, 2-bromo-1-2',4'-dimethoxyphenyl ethanone, 2-bromo-1-2,4-dimethoxyphenyl ethan-1-one, ethanone, 2-bromo-1-2,4-dimethoxyphenyl, 2-bromo-2',4'-dimethoxyacetopheneone, 1-2,4-dimethoxyphenyl-2-bromoethan-1-one, pubchem13433, acmc-1ba4l | |
| COC1=CC(=C(C=C1)C(=O)CBr)OC | |
| 259.099 | |
| 259.1 | |
| 2-Bromo-2',4'-dimethoxyacetophenone |
| 102°C to 105°C | |
| MFCD00000197 | |
| UN3261 | |
| PKVBZABQCCQHLD-UHFFFAOYSA-N | |
| 2-bromo-1-(2,4-dimethoxyphenyl)ethanone | |
| 98683 | |
| 98% |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber : 262-542-6
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only