missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-2',4'-dimethoxyacetophenone, 98%

Catalog No. AAB2230003
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2230003 1 g
AAB2230006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2230003 Supplier Thermo Scientific Chemicals Supplier No. B2230003

CAS: 60965-26-6 | C10H11BrO3 | 259.099 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 60965-26-6
Molecular Formula C10H11BrO3
Molecular Weight (g/mol) 259.099
MDL Number MFCD00000197
InChI Key PKVBZABQCCQHLD-UHFFFAOYSA-N
Synonym 2-bromo-1-2,4-dimethoxyphenyl ethanone, 2-bromo-2',4'-dimethoxyacetophenone, 2,4-dimethoxyphenacyl bromide, 2-bromo-1-2',4'-dimethoxyphenyl ethanone, 2-bromo-1-2,4-dimethoxyphenyl ethan-1-one, ethanone, 2-bromo-1-2,4-dimethoxyphenyl, 2-bromo-2',4'-dimethoxyacetopheneone, 1-2,4-dimethoxyphenyl-2-bromoethan-1-one, pubchem13433, acmc-1ba4l
PubChem CID 98683
IUPAC Name 2-bromo-1-(2,4-dimethoxyphenyl)ethanone
SMILES COC1=CC(=C(C=C1)C(=O)CBr)OC

Specifications

Melting Point 102°C to 105°C
Quantity 1 g
UN Number UN3261
Formula Weight 259.1
Percent Purity 98%
Chemical Name or Material 2-Bromo-2',4'-dimethoxyacetophenone
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 262-542-6
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only