missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-2',4'-difluoroacetophenone 98.0+%, TCI America™

Catalog No. p-7123303
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B40205G 5 g
B402025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B40205G Supplier TCI America Supplier No. B40205G
Only null left

Chemical Identifiers

CAS 102429-07-2
Molecular Formula C8H5BrF2O
Molecular Weight (g/mol) 235.028
MDL Number MFCD00142822
InChI Key CSGDTHXBRAAOHV-UHFFFAOYSA-N
Synonym 2-bromo-2',4'-difluoroacetophenone, 2-bromo-1-2,4-difluorophenyl ethanone, 2,4-difluorophenacyl bromide, 2-bromo-1-2,4-difluorophenyl ethan-1-one, 2-bromo-1-2,4-difluoro-phenyl-ethanone, 2',4'-difluorophenacyl bromide, 2-bromo-2,4-difluoroacetophenone, ethanone, 2-bromo-1-2,4-difluorophenyl, alpha-bromo-2',4'-difluoroacetophenone, acmc-1c4aw
PubChem CID 2774185
IUPAC Name 2-bromo-1-(2,4-difluorophenyl)ethanone
SMILES C1=CC(=C(C=C1F)F)C(=O)CBr

Specifications

Melting Point 35°C
Quantity 5 g
UN Number 2925
Formula Weight 235.03
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromo-2′,4′-difluoroacetophenone