missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-2',4'-difluoroacetophenone, 95%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 442280010

 View more versions of this product

Catalog No. AC442280010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

2-Bromo-2', 4'-difluoroacetophenone
94.0
51°C
94% min. (GC)
C8H5BrF2O
2-bromo-2',4'-difluoroacetophenone, 2-bromo-1-2,4-difluorophenyl ethanone, 2,4-difluorophenacyl bromide, 2-bromo-1-2,4-difluorophenyl ethan-1-one, 2-bromo-1-2,4-difluoro-phenyl-ethanone, 2',4'-difluorophenacyl bromide, 2-bromo-2,4-difluoroacetophenone, ethanone, 2-bromo-1-2,4-difluorophenyl, alpha-bromo-2',4'-difluoroacetophenone, acmc-1c4aw
C1=CC(=C(C=C1F)F)C(=O)CBr
235.03
235.03
102429-07-2
100.0
Authentic
Glass Bottle
1g
CSGDTHXBRAAOHV-UHFFFAOYSA-N
2-bromo-1-(2,4-difluorophenyl)ethanone
2774185
95%

Chemical Identifiers

102429-07-2
235.03
2-bromo-2',4'-difluoroacetophenone, 2-bromo-1-2,4-difluorophenyl ethanone, 2,4-difluorophenacyl bromide, 2-bromo-1-2,4-difluorophenyl ethan-1-one, 2-bromo-1-2,4-difluoro-phenyl-ethanone, 2',4'-difluorophenacyl bromide, 2-bromo-2,4-difluoroacetophenone, ethanone, 2-bromo-1-2,4-difluorophenyl, alpha-bromo-2',4'-difluoroacetophenone, acmc-1c4aw
2-bromo-1-(2,4-difluorophenyl)ethanone
C8H5BrF2O
CSGDTHXBRAAOHV-UHFFFAOYSA-N
2774185
C1=CC(=C(C=C1F)F)C(=O)CBr
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
Flammable solid.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Immediately call

GHS Signal Word: Danger

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only