missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-1H-imidazole, 98%

Catalog No. AC454630010
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC454630010 1 g
AC454630050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC454630010 Supplier Thermo Scientific Chemicals Supplier No. 454630010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 16681-56-4 | C3H3BrN2 | 146.98 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 16681-56-4
Molecular Formula C3H3BrN2
Molecular Weight (g/mol) 146.98
MDL Number MFCD02179526
InChI Key AXHRGVJWDJDYPO-UHFFFAOYSA-N
Synonym 2-bromoimidazole, 1h-imidazole, 2-bromo, bromoimidazole, sftheabililuh@, pubchem8987, zlchem 1065, acmc-1btox, 2-bromanyl-1h-imidazole, ksc176e6b, 2-bromo-1h-imidazole
PubChem CID 2773261
IUPAC Name 2-bromo-1H-imidazole
SMILES BrC1=NC=CN1

Specifications

Melting Point 189.0°C to 190.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 146.97
Percent Purity 98%
Chemical Name or Material 2-Bromo-1H-imidazole
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye damage.
May cause respiratory irritation.
Precautionary Statement GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. Immediately call a POISON CENTER or doctor/physician. IF ON SKIN: Wash with plenty of soap and water. IF INHALED: Call a POISON CENTER or doctor/physician if you feel unwell. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only