missing translation for 'onlineSavingsMsg'
Learn More

2-bromo-1-phenyl-1H-imidazole, Thermo Scientific™

Catalog No. CC75910DA
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC75910DA 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CC75910DA Supplier Thermo Scientific Chemicals Supplier No. CC75910DA

Chemical Identifiers

CAS 1034566-05-6
Molecular Formula C9H7BrN2
Molecular Weight (g/mol) 223.07
InChI Key KKPQUNLAAJJUCR-UHFFFAOYSA-N
Synonym 2-bromo-1-phenyl-1h-imidazole, 2-bromo-1-phenylimidazole
PubChem CID 45594301

Specifications

Packaging Amber glass bottle
Quantity 1 g
Formula Weight 223.07
Percent Purity 97%
Chemical Name or Material 2-bromo-1-phenyl-1H-imidazole
Hazard Category GHS Signal Word: Warning