missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-1-methylimidazole, 95%

Catalog No. p-7034710
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5240703 1 g
AAH5240706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
Catalog No. AAH5240703 missing translation for 'mfr' Thermo Scientific Chemicals missing translation for 'supplierNo' H5240703

CAS: 16681-59-7 | C4H5BrN2 | 161.002 g/mol

2-Bromo-1-methylimidazole is used in in pharmaceuticals, drug candidates, ligands for transition metal catalysts and other molecular functional materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromo-1-methylimidazole is used in in pharmaceuticals, drug candidates, ligands for transition metal catalysts and other molecular functional materials.

Notes
Stable under recommended storage conditions. Store away from oxidizing agents.

Chemical Identifiers

CAS 16681-59-7
Molecular Formula C4H5BrN2
Molecular Weight (g/mol) 161.002
MDL Number MFCD02179525
InChI Key BANOTGHIHYMTDL-UHFFFAOYSA-N
Synonym 2-bromo-1-methyl-1h-imidazole, 1-methyl-2-bromoimidazole, 2-bromo-n-methylimidazole, 1h-imidazole, 2-bromo-1-methyl, zlchem 620, pubchem7594, n-methyl-2-bromoimidazole, acmc-209dv3, 2-bromanyl-1-methyl-imidazole, 1-methyl-2-bromo-1h-imidazole
PubChem CID 2773262
IUPAC Name 2-bromo-1-methylimidazole
SMILES CN1C=CN=C1Br

Specifications

Density 1.649
Boiling Point 172°C
Flash Point >110°C (230°F)
Quantity 1 g
Formula Weight 161
Percent Purity 95%
Chemical Name or Material 2-Bromo-1-methylimidazole
missing translation for 'healthHazard1' H302-H315-H318
missing translation for 'healthHazard2' GHS H Statement
H301-H318-H315
Toxic if swallowed.
Causes serious eye damage.
Causes skin irritation.
missing translation for 'healthHazard3' P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P310-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only