missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromo-1-ethylpyridinium Tetrafluoroborate 98.0+%, TCI America™
$173.37 - $436.19
Chemical Identifiers
| CAS | 878-23-9 |
|---|---|
| Molecular Formula | C7H9BBrF4N |
| Molecular Weight (g/mol) | 273.86 |
| MDL Number | MFCD00060046 |
| InChI Key | YJDXVQLBIAJTHP-UHFFFAOYSA-N |
| Synonym | 2-bromo-1-ethylpyridinium tetrafluoroborate, 2-bromo-1-ethylpyridin-1-ium tetrafluoroborate, bep tetrafluoroborate, bromoethylpyridiniumtetrafluoroborate, 2-bromo-1-ethyl-pyridinium tetrafluoroborate, bepybr, acmc-209qqc, 2-brom-1-ethyl-pyridiniumtetrafluoroborat, 2-bromo-1-ethyl pyridinium tetrafluoroborate, 2-bromo-1-ethyl-pyridin-1-ium tetrafluoroborate |
| PubChem CID | 2736300 |
| IUPAC Name | 2-bromo-1-ethylpyridin-1-ium; tetrafluoroboranuide |
| SMILES | F[B-](F)(F)F.CC[N+]1=CC=CC=C1Br |
Chemical Identifiers
| 878-23-9 | |
| 273.86 | |
| YJDXVQLBIAJTHP-UHFFFAOYSA-N | |
| 2736300 | |
| F[B-](F)(F)F.CC[N+]1=CC=CC=C1Br |
| C7H9BBrF4N | |
| MFCD00060046 | |
| 2-bromo-1-ethylpyridinium tetrafluoroborate, 2-bromo-1-ethylpyridin-1-ium tetrafluoroborate, bep tetrafluoroborate, bromoethylpyridiniumtetrafluoroborate, 2-bromo-1-ethyl-pyridinium tetrafluoroborate, bepybr, acmc-209qqc, 2-brom-1-ethyl-pyridiniumtetrafluoroborat, 2-bromo-1-ethyl pyridinium tetrafluoroborate, 2-bromo-1-ethyl-pyridin-1-ium tetrafluoroborate | |
| 2-bromo-1-ethylpyridin-1-ium; tetrafluoroboranuide |
Specifications
| 878-23-9 | |
| White | |
| MFCD00060046 | |
| 1759 | |
| YJDXVQLBIAJTHP-UHFFFAOYSA-N | |
| 2-bromo-1-ethylpyridin-1-ium; tetrafluoroboranuide | |
| 2736300 | |
| ≥98.0% (HPLC,T) | |
| 2-Bromo-1-ethylpyridinium Tetrafluoroborate |
| 103°C | |
| C7H9BBrF4N | |
| 5 g | |
| 2-bromo-1-ethylpyridinium tetrafluoroborate, 2-bromo-1-ethylpyridin-1-ium tetrafluoroborate, bep tetrafluoroborate, bromoethylpyridiniumtetrafluoroborate, 2-bromo-1-ethyl-pyridinium tetrafluoroborate, bepybr, acmc-209qqc, 2-brom-1-ethyl-pyridiniumtetrafluoroborat, 2-bromo-1-ethyl pyridinium tetrafluoroborate, 2-bromo-1-ethyl-pyridin-1-ium tetrafluoroborate | |
| F[B-](F)(F)F.CC[N+]1=CC=CC=C1Br | |
| 273.86 | |
| 273.86 | |
| Crystalline Powder |
Safety and Handling
TSCA : No