missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-1-(5-phenyl-2-thienyl)-1-ethanone, 95%, Thermo Scientific™

Catalog No. p-2697873
Change view
Click to view available options
Quantity:
250mg
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC10505CB 250mg
CC10505DA 1g
CC10505DE 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CC10505CB Supplier Thermo Scientific Chemicals Supplier No. CC10505CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 10531-43-8
Molecular Formula C12H9BrOS
Molecular Weight (g/mol) 281.167
MDL Number MFCD02681997
InChI Key BOIAFAJKOBVLGS-UHFFFAOYSA-N
Synonym 2-bromo-1-5-phenyl-2-thienyl-1-ethanone, 2-bromo-1-5-phenylthiophen-2-yl ethanone, ethanone,2-bromo-1-5-phenyl-2-thienyl, ethanone, 2-bromo-1-5-phenyl-2-thienyl, 2-bromoacetyl-5-phenylthiophene, bromomethyl 5-phenyl-2-thienyl ketone, 2-bromo-1-5-phenyl-2-thienyl-ethanone, 2-bromo-1-5-phenyl-2-thiophenyl ethanone, 2-bromo-1-5-phenylthien-2-yl ethan-1-one
PubChem CID 2776310
IUPAC Name 2-bromo-1-(5-phenylthiophen-2-yl)ethanone
SMILES C1=CC=C(C=C1)C2=CC=C(S2)C(=O)CBr

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber glass bottle
Quantity 250mg
Formula Weight 281.17
Percent Purity 95%
Chemical Name or Material 2-Bromo-1-(5-phenyl-2-thienyl)-1-ethanone
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.