missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-1-(5-phenyl-2-thienyl)-1-ethanone, 95%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC10505DE

Quantity 5g

 View more versions of this product

Catalog No. CC10505DE

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

10531-43-8
281.167
BOIAFAJKOBVLGS-UHFFFAOYSA-N
2776310
C1=CC=C(C=C1)C2=CC=C(S2)C(=O)CBr
C12H9BrOS
MFCD02681997
2-bromo-1-5-phenyl-2-thienyl-1-ethanone, 2-bromo-1-5-phenylthiophen-2-yl ethanone, ethanone,2-bromo-1-5-phenyl-2-thienyl, ethanone, 2-bromo-1-5-phenyl-2-thienyl, 2-bromoacetyl-5-phenylthiophene, bromomethyl 5-phenyl-2-thienyl ketone, 2-bromo-1-5-phenyl-2-thienyl-ethanone, 2-bromo-1-5-phenyl-2-thiophenyl ethanone, 2-bromo-1-5-phenylthien-2-yl ethan-1-one
2-bromo-1-(5-phenylthiophen-2-yl)ethanone

Specifications

2-Bromo-1-(5-phenyl-2-thienyl)-1-ethanone
94.5
95%
5g
2-bromo-1-5-phenyl-2-thienyl-1-ethanone, 2-bromo-1-5-phenylthiophen-2-yl ethanone, ethanone,2-bromo-1-5-phenyl-2-thienyl, ethanone, 2-bromo-1-5-phenyl-2-thienyl, 2-bromoacetyl-5-phenylthiophene, bromomethyl 5-phenyl-2-thienyl ketone, 2-bromo-1-5-phenyl-2-thienyl-ethanone, 2-bromo-1-5-phenyl-2-thiophenyl ethanone, 2-bromo-1-5-phenylthien-2-yl ethan-1-one
C1=CC=C(C=C1)C2=CC=C(S2)C(=O)CBr
281.167
281.17
10531-43-8
100.0
C12H9BrOS
MFCD02681997
BOIAFAJKOBVLGS-UHFFFAOYSA-N
2-bromo-1-(5-phenylthiophen-2-yl)ethanone
2776310
95%
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
Documents

Documents