missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-1-(4-methyl-2-phenyl-1,3-thiazol-5-yl)-1-ethanone, 95%, Thermo Scientific™

$301.51 - $301.51

Chemical Identifiers

CAS 7520-95-8
Molecular Formula C12H10BrNOS
Molecular Weight (g/mol) 296.182
MDL Number MFCD04071443
InChI Key BOMSILGSXFNLJX-UHFFFAOYSA-N
Synonym 2-bromo-1-4-methyl-2-phenyl-1,3-thiazol-5-yl-1-ethanone, 2-bromo-1-4-methyl-2-phenyl-1,3-thiazol-5-yl ethanone, 5-bromoacetyl-4-methyl-2-phenylthiazole, 2-bromo-1-4-methyl-2-phenylthiazol-5-yl ethanone, 2-bromo-1-4-methyl-2-phenyl 1,3-thiazol-5-yl ethan-1-one
PubChem CID 2795492
IUPAC Name 2-bromo-1-(4-methyl-2-phenyl-1,3-thiazol-5-yl)ethanone
SMILES CC1=C(SC(=N1)C2=CC=CC=C2)C(=O)CBr
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
CC29205DA
SDS View Documents
Thermo Scientific Chemicals
CC29205DA
1 g
Each for $301.51
Only null left
 
Description

Description

Specifications

Chemical Identifiers

7520-95-8
296.182
BOMSILGSXFNLJX-UHFFFAOYSA-N
2795492
CC1=C(SC(=N1)C2=CC=CC=C2)C(=O)CBr
C12H10BrNOS
MFCD04071443
2-bromo-1-4-methyl-2-phenyl-1,3-thiazol-5-yl-1-ethanone, 2-bromo-1-4-methyl-2-phenyl-1,3-thiazol-5-yl ethanone, 5-bromoacetyl-4-methyl-2-phenylthiazole, 2-bromo-1-4-methyl-2-phenylthiazol-5-yl ethanone, 2-bromo-1-4-methyl-2-phenyl 1,3-thiazol-5-yl ethan-1-one
2-bromo-1-(4-methyl-2-phenyl-1,3-thiazol-5-yl)ethanone

Specifications

7520-95-8
100.0
Amber glass bottle
MFCD04071443
2-bromo-1-4-methyl-2-phenyl-1,3-thiazol-5-yl-1-ethanone, 2-bromo-1-4-methyl-2-phenyl-1,3-thiazol-5-yl ethanone, 5-bromoacetyl-4-methyl-2-phenylthiazole, 2-bromo-1-4-methyl-2-phenylthiazol-5-yl ethanone, 2-bromo-1-4-methyl-2-phenyl 1,3-thiazol-5-yl ethan-1-one
CC1=C(SC(=N1)C2=CC=CC=C2)C(=O)CBr
296.182
296.19
2-Bromo-1-(4-methyl-2-phenyl-1, 3-thiazol-5-yl)-1-ethanone
89.5
90%
C12H10BrNOS
1 g
BOMSILGSXFNLJX-UHFFFAOYSA-N
2-bromo-1-(4-methyl-2-phenyl-1,3-thiazol-5-yl)ethanone
2795492
90%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS
missing translation for 'documents'

missing translation for 'documents'