missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-1-[4-(2-thienyl)phenyl]-1-ethanone, ≥97%, Thermo Scientific™

Catalog No. p-3110259
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC12005DA 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CC12005DA Supplier Thermo Scientific Chemicals Supplier No. CC12005DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 128746-80-5
Molecular Formula C12H9BrOS
Molecular Weight (g/mol) 281.167
MDL Number MFCD06659059
InChI Key QFHKNYPNLSJEQI-UHFFFAOYSA-N
PubChem CID 2795250
IUPAC Name 2-bromo-1-(4-thiophen-2-ylphenyl)ethanone
SMILES C1=CSC(=C1)C2=CC=C(C=C2)C(=O)CBr

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber glass bottle
Quantity 1 g
Synonym 2-bromo-1-4-2-thienyl phenyl-1-ethanone, ethanone,2-bromo-1-4-2-thienyl phenyl, 2-bromo-1-4-thiophen-2-yl phenyl ethanone, acmc-20msyh, 2-bromo-1-thien-2yphenyl ethanone, 2-bromo-4'-thien-2-yl acetophenone, 2-bromo-1-4-thiophen-2-ylphenyl ethanone, ethanone, 2-bromo-1-4-2-thienyl phenyl, 2-bromo-1-4-thien-2-yl phenyl ethan-1-one
Formula Weight 281.17
Percent Purity 97%
Chemical Name or Material 2-Bromo-1-[4-(2-thienyl)phenyl]-1-ethanone
Hazard Statement GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.
Harmful in contact with skin.
Causes serious eye damage.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.