missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-1-[4-(2-furyl)phenyl]ethanone, Tech., Thermo Scientific™

Catalog No. CC34405CB
Change view
Click to view available options
Quantity:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC34405CB 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CC34405CB Supplier Thermo Scientific Chemicals Supplier No. CC34405CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 868755-47-9
Molecular Formula C12H9BrO2
Molecular Weight (g/mol) 265.106
MDL Number MFCD08435854
InChI Key SONOLDBWXMKWPY-UHFFFAOYSA-N
Synonym 2-bromo-1-4-2-furyl phenyl ethanone, 2-bromo-1-4-furan-2-yl phenyl ethanone, 2-bromo-1-4-2-furyl phenyl ethan-1-one, ethanone,2-bromo-1-4-2-furanyl phenyl, 2-bromo-1-4-furan-2-yl phenyl ethan-1-one
PubChem CID 18525721
IUPAC Name 2-bromo-1-[4-(furan-2-yl)phenyl]ethanone
SMILES C1=COC(=C1)C2=CC=C(C=C2)C(=O)CBr

Specifications

CAS Min % 80.0
CAS Max % 100.0
Linear Formula Yes
Quantity 250mg
Formula Weight 265.11
Percent Purity Tech.
Grade Technical
Chemical Name or Material 2-Bromo-1-[4-(2-furyl)phenyl]ethanone
Hazard Statement GHS H Statement:
Harmful in contact with skin.
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful if inhaled.
Causes serious eye damage.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.