missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-1-(3-thienyl)-1-ethanone, 97%, Thermo Scientific™

Catalog No. p-4510104
Change view
Click to view available options
Quantity:
5g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC13105DE 5g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CC13105DE Supplier Thermo Scientific Chemicals Supplier No. CC13105DE

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1468-82-2
Molecular Formula C6H5BrOS
Molecular Weight (g/mol) 205.07
MDL Number MFCD02677695
InChI Key TXEJYUFJFSPCHH-UHFFFAOYSA-N
Synonym 2-bromo-1-3-thienyl-1-ethanone, 2-bromo-1-thiophen-3-yl ethanone, 3-bromoacetylthiophene, 3-bromoacetyl thiophene, 2-bromo-1-thiophen-3-yl ethan-1-one, 2-bromo-1-3-thienyl ethanone, ethanone,2-bromo-1-3-thienyl, ethanone, 2-bromo-1-3-thienyl, 2-bromo-1-thien-3-yl-1-ethanone
PubChem CID 2776379
IUPAC Name 2-bromo-1-thiophen-3-ylethanone
SMILES BrCC(=O)C1=CSC=C1

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 5g
Formula Weight 205.07
Percent Purity 97%
Chemical Name or Material 2-Bromo-1-(3-thienyl)-1-ethanone