missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-1-(3-chloro-4-methylphenyl)propan-1-one, Tech., Thermo Scientific™

Catalog No. p-2697790
Change view
Click to view available options
Quantity:
1g
10g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
BTB09098DA 1g Amber Glass Bottle
BTB09098EA 10g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BTB09098DA Supplier Thermo Scientific Chemicals Supplier No. BTB09098DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 175135-93-0
Molecular Formula C10H10BrClO
Molecular Weight (g/mol) 261.543
MDL Number MFCD00067862
InChI Key RGMFWCVOMGKIJM-UHFFFAOYSA-N
Synonym 2-bromo-1-3-chloro-4-methylphenyl propan-1-one, 2-bromo-3'-chloro-4'-methylpropiophenone, 1-propanone,2-bromo-1-3-chloro-4-methylphenyl, 2-bromo-3'-chloro-4'methyl propiophenone, 2-bromo-1-3-chloro-4-methylphenyl-1-propanone, 2-bromanyl-1-3-chloranyl-4-methyl-phenyl propan-1-one
PubChem CID 2774950
IUPAC Name 2-bromo-1-(3-chloro-4-methylphenyl)propan-1-one
SMILES CC1=C(C=C(C=C1)C(=O)C(C)Br)Cl

Specifications

CAS Min % 80.0
CAS Max % 100.0
Packaging Amber Glass Bottle
Quantity 1g
Formula Weight 261.55
Percent Purity Tech.
Grade Technical
Chemical Name or Material 2-Bromo-1-(3-chloro-4-methylphenyl)propan-1-one
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.