missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-1,3,5-trichlorobenzene, 97%

Catalog No. AAB2500006
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2500006 5 g
AAB2500014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2500006 Supplier Thermo Scientific Chemicals Supplier No. B2500006

CAS: 19393-96-5 | C6H2BrCl3 | 260.34 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 19393-96-5
Molecular Formula C6H2BrCl3
Molecular Weight (g/mol) 260.34
MDL Number MFCD00060653
InChI Key BAPPAFMEUDJAQI-UHFFFAOYSA-N
PubChem CID 140526
IUPAC Name 2-bromo-1,3,5-trichlorobenzene
SMILES ClC1=CC(Cl)=C(Br)C(Cl)=C1

Specifications

Quantity 5 g
Synonym 1-bromo-2,4,6-trichlorobenzene, 2,4,6-trichlorobromobenzene, benzene, 2-bromo-1,3,5-trichloro, benzene,2-bromo-1,3,5-trichloro, acmc-20aoxf, 1,3,5-trichloro-2-bromobenzene, 2,4,6-trichloro-1-bromobenzene, 2-bromo-1,3,5-trichloro-benzene, #, 2-bromanyl-1,3,5-tris chloranyl benzene
Formula Weight 260.34
Percent Purity 97%
Chemical Name or Material 2-Bromo-1,3,5-trichlorobenzene
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only