missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromo-1-(2-pyridinyl)-1-ethanone hydrobromide, Thermo Scientific™
Specifications
Specifications
| Assay Percent Range | 90% |
| Packaging | Amber Glass Bottle |
| Quantity | 1g |
| Formula Weight | 280.95 |
| Percent Purity | 90% |
| Chemical Name or Material | 2-Bromo-1-(2-pyridinyl)-1-ethanone hydrobromide |