missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromo-1-(2,4-dimethylphenyl)ethan-1-one, Thermo Scientific™
Specifications
Specifications
| Assay Percent Range | 97% |
| Packaging | Amber Glass Bottle |
| Linear Formula | (CH3)2C6H3C(O)CH2Br |
| Quantity | 1g |
| Formula Weight | 227.1 |
| Percent Purity | 97% |
| Chemical Name or Material | 2-Bromo-1-(2, 4-dimethylphenyl)ethan-1-one |