missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromo-1,10-phenanthroline 98.0+%, TCI America™
$128.62 - $380.27
Chemical Identifiers
| CAS | 22426-14-8 |
|---|---|
| Molecular Formula | C12H7BrN2 |
| Molecular Weight (g/mol) | 259.106 |
| MDL Number | MFCD08276287 |
| InChI Key | ZRJUDAZGVGIDLP-UHFFFAOYSA-N |
| PubChem CID | 12830247 |
| IUPAC Name | 2-bromo-1,10-phenanthroline |
| SMILES | C1=CC2=C(C3=C(C=C2)C=CC(=N3)Br)N=C1 |
Chemical Identifiers
| 22426-14-8 | |
| 259.106 | |
| ZRJUDAZGVGIDLP-UHFFFAOYSA-N | |
| 2-bromo-1,10-phenanthroline |
| C12H7BrN2 | |
| MFCD08276287 | |
| 12830247 | |
| C1=CC2=C(C3=C(C=C2)C=CC(=N3)Br)N=C1 |
Specifications
| 22426-14-8 | |
| C12H7BrN2 | |
| 1 g | |
| C1=CC2=C(C3=C(C=C2)C=CC(=N3)Br)N=C1 | |
| 259.106 | |
| 259.11 | |
| Crystalline Powder |
| 163°C | |
| MFCD08276287 | |
| ZRJUDAZGVGIDLP-UHFFFAOYSA-N | |
| 2-bromo-1,10-phenanthroline | |
| 12830247 | |
| ≥98.0% (T) | |
| 2-Bromo-1,10-phenanthroline |
Safety and Handling
TSCA : No