missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-1-(1-methyl-1H-benzimidazol-5-yl)ethanone hydrobromide, 97%, Thermo Scientific™

Catalog No. p-3586654
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC13655DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CC13655DA Supplier Thermo Scientific Chemicals Supplier No. CC13655DA

Chemical Identifiers

CAS 944450-78-6
Molecular Formula C10H10Br2N2O
Molecular Weight (g/mol) 334.011
MDL Number MFCD09817461
InChI Key LRWGKOQDEBXXPS-UHFFFAOYSA-N
Synonym 2-bromo-1-1-methyl-1h-benzimidazol-5-yl ethanone hydrobromide, 2-bromo-1-1-methyl-1h-benzo d imidazol-5-yl ethanone hydrobromide, 2-bromo-1-1-methyl-1h-benzimidazol-5-yl ethan-1-one hydrobromide, 2-bromo-1-1-methyl-1,3-benzodiazol-5-yl ethanone hydrobromide, c10h9brn2o.hbr, 2-bromo-1-1-methyl-1h-benzoimidazol-5-yl-ethanone.hbr, 2-bromo-1-1-methyl-1h-benzo d imidazol-5-yl ethan-1-one hydrobromide, 2-bromo-1-1-methyl-1h-benzimidazol-5-yl ethan-1-one-hydrogen bromide 1/1
PubChem CID 24229460
IUPAC Name 2-bromo-1-(1-methylbenzimidazol-5-yl)ethanone;hydrobromide
SMILES CN1C=NC2=C1C=CC(=C2)C(=O)CBr.Br

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 334.01
Percent Purity 97%
Chemical Name or Material 2-Bromo-1-(1-methyl-1H-benzimidazol-5-yl)ethanone hydrobromide
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Harmful in contact with skin.
Causes serious eye irritation.
Harmful if inhaled.
Harmful if swallowed.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.