missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-1-(1-methyl-1h-benzimidazol-2-yl)-1-ethanone, 97%, Thermo Scientific™

Catalog No. CC03805DA
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC03805DA 1 g Amber Glass Bottle
CC03805CB 250 mg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC03805DA Supplier Thermo Scientific Chemicals Supplier No. CC03805DA

Chemical Identifiers

CAS 56653-43-1
Molecular Formula C10H9BrN2O
Molecular Weight (g/mol) 253.10
MDL Number MFCD06200854
InChI Key KHFRWYRANOMZCP-UHFFFAOYSA-N
Synonym 2-bromo-1-1-methyl-1h-benzimidazol-2-yl-1-ethanone, 2-bromo-1-1-methyl-1,3-benzodiazol-2-yl ethanone, 2-bromo-1-1-methyl-1h-benzo d imidazol-2-yl-1-ethanone, 1-methyl-2-bromoacetyl-1h-benzimidazole, 2-bromo-1-1-methylbenzimidazol-2-yl ethanone, 2-bromo-1-1-methyl-1h-benzimidazol-2-yl ethanone, 2-bromo-1-1-methylbenzimidazol-2-yl ethan-1-one, 2-bromo-1-1-methyl-1h-benzoimidazol-2-yl-ethanone, 2-bromo-1-1-methyl-1h-benzoimidazol-2-yl ethanone, 2-bromo-1-1-methyl-1h-benzo d imidazol-2-yl ethanone
PubChem CID 2795119
IUPAC Name 2-bromo-1-(1-methylbenzimidazol-2-yl)ethanone
SMILES CN1C(=NC2=CC=CC=C12)C(=O)CBr

Specifications

Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 253.1
Percent Purity 97%
Chemical Name or Material 2-Bromo-1-(1-methyl-1H-benzimidazol-2-yl)-1-ethanone