missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(Boc-amino)thiazole, 97%
CAS: 170961-15-6 | C8H12N2O2S | 200.256 g/mol
$295.96 - $1133.36
Chemical Identifiers
| CAS | 170961-15-6 |
|---|---|
| Molecular Formula | C8H12N2O2S |
| Molecular Weight (g/mol) | 200.256 |
| MDL Number | MFCD09038327 |
| InChI Key | NCJXQSNROJRSSL-UHFFFAOYSA-N |
| Synonym | tert-butyl thiazol-2-ylcarbamate, tert-butyl n-1,3-thiazol-2-yl carbamate, n-boc-2-aminothiazole, 2-boc-amino thiazole, 2-tert-butoxycarbonylamino thiazole, thiazol-2-yl-carbamic acid tert-butyl ester, pubchem23669, tertbutylthiazolylcarbamate, 2-n-boc-amino thiazole |
| PubChem CID | 11858898 |
| IUPAC Name | tert-butyl N-(1,3-thiazol-2-yl)carbamate |
| SMILES | CC(C)(C)OC(=O)NC1=NC=CS1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH5856106
|
Thermo Scientific Chemicals
H5856106 |
5 g |
Each for $295.96
|
|
|||||
|
AAH5856115
|
Thermo Scientific Chemicals
H5856115 |
30 g |
Each for $1,133.36
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 170961-15-6 | |
| 200.256 | |
| NCJXQSNROJRSSL-UHFFFAOYSA-N | |
| 11858898 | |
| CC(C)(C)OC(=O)NC1=NC=CS1 |
| C8H12N2O2S | |
| MFCD09038327 | |
| tert-butyl thiazol-2-ylcarbamate, tert-butyl n-1,3-thiazol-2-yl carbamate, n-boc-2-aminothiazole, 2-boc-amino thiazole, 2-tert-butoxycarbonylamino thiazole, thiazol-2-yl-carbamic acid tert-butyl ester, pubchem23669, tertbutylthiazolylcarbamate, 2-n-boc-amino thiazole | |
| tert-butyl N-(1,3-thiazol-2-yl)carbamate |
Specifications
| 180°C to 185°C | |
| C8H12N2O2S | |
| tert-butyl thiazol-2-ylcarbamate, tert-butyl n-1,3-thiazol-2-yl carbamate, n-boc-2-aminothiazole, 2-boc-amino thiazole, 2-tert-butoxycarbonylamino thiazole, thiazol-2-yl-carbamic acid tert-butyl ester, pubchem23669, tertbutylthiazolylcarbamate, 2-n-boc-amino thiazole | |
| CC(C)(C)OC(=O)NC1=NC=CS1 | |
| 5 g | |
| 11858898 | |
| 97% |
| 170961-15-6 | |
| MFCD09038327 | |
| NCJXQSNROJRSSL-UHFFFAOYSA-N | |
| tert-butyl N-(1,3-thiazol-2-yl)carbamate | |
| 200.256 | |
| 200.25 | |
| 2-(Boc-amino)thiazole |
Safety and Handling
GHS H Statement
H301-H317
Toxic if swallowed.
May cause an allergic skin reaction.
P261-P264b-P270-P272-P280g-P301+P312-P302+P352-P330-P333+P313-P363-P501c
H302-H317
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only