missing translation for 'onlineSavingsMsg'
Learn More

2-(Boc-amino)thiazole, 97%

Catalog No. AAH5856115
Change view
Click to view available options
Quantity:
5 g
30 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5856115 30 g
AAH5856106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5856115 Supplier Thermo Scientific Chemicals Supplier No. H5856115
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 170961-15-6 | C8H12N2O2S | 200.256 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 170961-15-6
Molecular Formula C8H12N2O2S
Molecular Weight (g/mol) 200.256
MDL Number MFCD09038327
InChI Key NCJXQSNROJRSSL-UHFFFAOYSA-N
Synonym tert-butyl thiazol-2-ylcarbamate, tert-butyl n-1,3-thiazol-2-yl carbamate, n-boc-2-aminothiazole, 2-boc-amino thiazole, 2-tert-butoxycarbonylamino thiazole, thiazol-2-yl-carbamic acid tert-butyl ester, pubchem23669, tertbutylthiazolylcarbamate, 2-n-boc-amino thiazole
PubChem CID 11858898
IUPAC Name tert-butyl N-(1,3-thiazol-2-yl)carbamate
SMILES CC(C)(C)OC(=O)NC1=NC=CS1

Specifications

Melting Point 180°C to 185°C
Quantity 30 g
Formula Weight 200.25
Percent Purity 97%
Chemical Name or Material 2-(Boc-amino)thiazole
Hazard Category H302-H317
Hazard Statement GHS H Statement
H301-H317
Toxic if swallowed.
May cause an allergic skin reaction.
Precautionary Statement P261-P264b-P270-P272-P280g-P301+P312-P302+P352-P330-P333+P313-P363-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only