missing translation for 'onlineSavingsMsg'
Learn More

2-(Boc-amino)pyridine, 95%

Catalog No. AAH3074103
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3074103 1 g
AAH3074106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3074103 Supplier Thermo Scientific Chemicals Supplier No. H3074103
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 38427-94-0 | C10H14N2O2 | 194.234 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 38427-94-0
Molecular Formula C10H14N2O2
Molecular Weight (g/mol) 194.234
MDL Number MFCD03411622
InChI Key ORUGTGTZBRUQIT-UHFFFAOYSA-N
Synonym 2-boc-amino pyridine, tert-butyl pyridin-2-ylcarbamate, 2-tert-butoxycarbonylamino pyridine, tert-butyl n-pyridin-2-yl carbamate, tert-butyl n-2-pyridyl carbamate, pyridin-2-yl-carbamic acid tert-butyl ester, carbamic acid, 2-pyridinyl-, 1,1-dimethylethyl ester, carbamic acid, n-2-pyridinyl-, 1,1-dimethylethyl ester, pubchem17730, boc-2-aminopyridine
PubChem CID 11206349
IUPAC Name tert-butyl N-pyridin-2-ylcarbamate
SMILES CC(C)(C)OC(=O)NC1=CC=CC=N1

Specifications

Melting Point 92°C to 94°C
Quantity 1 g
Formula Weight 194.23
Percent Purity 95%
Chemical Name or Material 2-(Boc-amino)pyridine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only