missing translation for 'onlineSavingsMsg'
Learn More

2-(Boc-amino)pyridine-5-carboxaldehyde, 97%, Thermo Scientific Chemicals

Catalog No. AAH5007503
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5007503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH5007503 Supplier Thermo Scientific Chemicals Supplier No. H5007503
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 199296-40-7 | Molecular Formula: C11H14N2O3 | Molecular Weight: 222.244 g/mol

2-(Boc-amino)pyridine-5-carboxaldehyde is used as active pharmaceutical ingredient or intermediates, fine chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(Boc-amino)pyridine-5-carboxaldehyde is used as active pharmaceutical ingredient or intermediates, fine chemicals.

Solubility
Soluble in water.

Notes
Air & Light Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 199296-40-7
Molecular Formula C11H14N2O3
Molecular Weight (g/mol) 222.244
MDL Number MFCD08064228
InChI Key WZROBBWIJBBWQP-UHFFFAOYSA-N
Synonym tert-butyl 5-formylpyridin-2-yl carbamate, tert-butyl 5-formylpyridin-2-ylcarbamate, tert-butyl n-5-formylpyridin-2-yl carbamate, 2-boc-amino pyridine-5-carboxaldehyde, 5-formyl-pyridin-2-yl-carbamic acid tert-butyl ester, 5-formylpyridin-2-yl carbamic acid tert butyl ester, 2-boc-aminopyridine-5-aldehyde, 2-tert-butoxycarbonylamino pyridine-5-carboxaldehyde, carbamicacid, n-5-formyl-2-pyridinyl-, 1,1-dimethylethyl ester, 6-aminonicotinaldehyde, 6-boc protected
PubChem CID 22034247
IUPAC Name tert-butyl N-(5-formylpyridin-2-yl)carbamate
SMILES CC(C)(C)OC(=O)NC1=NC=C(C=C1)C=O

Specifications

Quantity 1 g
Sensitivity Air and light sensitive
Solubility Information Soluble in water.
Formula Weight 222.25
Percent Purity 97%
Chemical Name or Material 2-(Boc-amino)pyridine-5-carboxaldehyde
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only