missing translation for 'onlineSavingsMsg'
Learn More

2-(Boc-amino)pyridine-3-boronic acid pinacol ester, 98%, Thermo Scientific Chemicals

Catalog No. AAH5009403
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5009403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH5009403 Supplier Thermo Scientific Chemicals Supplier No. H5009403
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1072944-99-0 | Molecular Formula: C16H25BN2O4 | Molecular Weight: 320.196 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Notes
Heat and light sensitive. Incompatible with oxidizing agents and strong reducing agents.

Chemical Identifiers

CAS 1072944-99-0
Molecular Formula C16H25BN2O4
Molecular Weight (g/mol) 320.196
MDL Number MFCD08063072
InChI Key FYKIZSWVZIHFLF-UHFFFAOYSA-N
Synonym 2-boc-amino pyridine-3-boronic acid pinacol ester, tert-butyl 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridin-2-ylcarbamate, tert-butyl n-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridin-2-yl carbamate, 2-tert-butoxycarbonylamino pyridine-3-boronic acid pinacol ester, tert-butyl 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridin-2-yl carbamate, 2-n-boc-amino pyridine-3-boronic acid, pinacol ester, tert-butyl 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2-pyridyl carbamate, 3-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-pyridin-2-yl-carbamic acid tert-butyl ester, tert-butyl 3-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl pyridin-2-yl carbamate
PubChem CID 45925650
IUPAC Name tert-butyl N-[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-yl]carbamate
SMILES B1(OC(C(O1)(C)C)(C)C)C2=C(N=CC=C2)NC(=O)OC(C)(C)C

Specifications

Quantity 1 g
Sensitivity Air Sensitive
Formula Weight 320.2
Percent Purity 98%
Chemical Name or Material 2-(Boc-amino)pyridine-3-boronic acid pinacol ester
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only