missing translation for 'onlineSavingsMsg'
Learn More

2-(BOC-amino)ethyl bromide, 96%

Catalog No. AC398940050
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC398940050 5 g Glass Bottle
AC398940010 1 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC398940050 Supplier Thermo Scientific Chemicals Supplier No. 398940050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 39684-80-5 | C7H14BrNO2 | 224.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 39684-80-5
Molecular Formula C7H14BrNO2
Molecular Weight (g/mol) 224.10
MDL Number MFCD02683428
InChI Key TZRQZPMQUXEZMC-UHFFFAOYSA-N
Synonym tert-butyl n-2-bromoethyl carbamate, 2-boc-amino ethyl bromide, n-boc-bromoethylamine, tert-butyl 2-bromoethyl carbamate, n-boc-2-bromoethylamine, tert-butyl 2-bromoethylcarbamate, 2-bromo-ethyl-carbamic acid tert-butyl ester, 2-tert-butoxycarbonylamino ethyl bromide, 2-boc-amino-ethyl bromide
PubChem CID 4103526
IUPAC Name tert-butyl N-(2-bromoethyl)carbamate
SMILES CC(C)(C)OC(=O)NCCBr

Specifications

Melting Point 30°C to 32°C
CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass Bottle
Linear Formula (CH3)3COC(O)NHCH2CH2Br
Quantity 5 g
Formula Weight 224.09
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material 2-(BOC-amino)ethyl bromide
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only