missing translation for 'onlineSavingsMsg'
Learn More

2-Boc-2-azabicyclo[2.2.1]hept-5-ene, 98%

Catalog No. AAH6369806
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6369806 5 g
AAH6369803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6369806 Supplier Thermo Scientific Chemicals Supplier No. H6369806
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 188345-71-3 | C11H17NO2 | 195.262 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 188345-71-3
Molecular Formula C11H17NO2
Molecular Weight (g/mol) 195.262
MDL Number MFCD22416354
InChI Key ZATXHTCUZAWODK-UHFFFAOYSA-N
PubChem CID 15837921
IUPAC Name tert-butyl 5-azabicyclo[2.2.1]hept-2-ene-5-carboxylate
SMILES CC(C)(C)OC(=O)N1CC2CC1C=C2

Specifications

Color Yellow
Quantity 5 g
Synonym 2-boc-2-azabicyclo 2.2.1 hept-5-ene, tert-butyl 2-aza-bicyclo 2.2.1 hept-5-ene-2-carboxylate, tert-butyl 2-azabicyclo 2.2.1 hept-5-ene-2-carboxylate, 2-tert-butoxycarbonyl-2-azabicyclo 2.2.1 hept-5-ene, tert-butyl2-azabicyclo 2.2.1 hept-5-ene-2-carboxylate, ksc497c1n, tert-butyl 5-azabicyclo 2.2.1 hept-2-ene-5-carboxylate, 2-aza-bicyclo 2.2.1 hept-5-ene-2-carboxylic acid tert-butyl ester, 2-tert-butyloxycarbonyl-2-azabicyclo 2.2.1 hepta-5-ene, 2-azabicyclo 2.2.1 hept-5-ene-2-carboxylic acid tert-butyl ester
Formula Weight 195.26
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2-Boc-2-azabicyclo[2.2.1]hept-5-ene
Hazard Category H302-H317-H318
Hazard Statement GHS H Statement
H318-H302-H317
Causes serious eye damage.
Harmful if swallowed.
May cause an allergic skin reaction.
Precautionary Statement P261-P264b-P270-P272-P280-P301+P312-P302+P352-P305+P351+P338-P310-P330-P333+P313-P363-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only