missing translation for 'onlineSavingsMsg'
Learn More

2-Biphenylcarboxylic acid, 98%, Thermo Scientific™

Catalog No. AC106270250
Change view
Click to view available options
Quantity:
25g
100g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC106270250 25g Glass bottle
AC106271000 100g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC106270250 Supplier Thermo Scientific Chemicals Supplier No. 106270250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 947-84-2
Molecular Formula C13H10O2
Molecular Weight (g/mol) 198.22
MDL Number MFCD00002463
InChI Key ILYSAKHOYBPSPC-UHFFFAOYSA-N
Synonym 2-biphenylcarboxylic acid, 1,1'-biphenyl-2-carboxylic acid, o-phenylbenzoic acid, biphenyl-2-carboxylic acid, biphenylcarboxylic acid, 1,1'-biphenyl carboxylic acid, diphenyl-2-carboxylic acid, phenylbenzoic acid
PubChem CID 70357
IUPAC Name 2-phenylbenzoic acid
SMILES C1=CC=C(C=C1)C2=CC=CC=C2C(=O)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Boiling Point 199.0°C (10.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Linear Formula C6H5C6H4CO2H
Quantity 25g
Beilstein 09,669
Solubility Information Solubility in water: insoluble.
Formula Weight 198.22
Percent Purity 98%
Chemical Name or Material 2-Biphenylcarboxylic acid, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 213-432-1
TSCA TSCA

RUO – Research Use Only