missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Benzylpyridine, 98+%
Description
2-Benzylpyridine is used in organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Benzylpyridine is used in organic synthesis.
Solubility
Soluble in alcohol. Insoluble in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
2-Benzylpyridine is used in organic synthesis.
Solubility
Soluble in alcohol. Insoluble in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications
Specifications
| Melting Point | 10°C to 12°C |
| Density | 1.054 |
| Boiling Point | 140°C to 143°C (11 mmHg) |
| Flash Point | 125°C (257°F) |
| Refractive Index | 1.579 |
| Quantity | 250 g |
| Beilstein | 115875 |
| Solubility Information | Soluble in alcohol. Insoluble in water. |
| Formula Weight | 169.23 |
| Percent Purity | ≥98% |
| Show More |
Safety and Handling
| Hazard Category | H315-H319-H335 |
| Hazard Statement | GHS H StatementH315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
| EINECSNumber | 202-979-1 |
| RTECSNumber | US2620000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only