missing translation for 'onlineSavingsMsg'
Learn More

2-Benzylpyridine, 98+%

Catalog No. AAA1066218
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1066218 50 g
AAA1066230 250 g
AAA106620B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1066218 Supplier Thermo Scientific Chemicals Supplier No. A1066218
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 101-82-6 | C12H11N | 169.23 g/mol

2-Benzylpyridine is used in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Benzylpyridine is used in organic synthesis.

Solubility
Soluble in alcohol. Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 101-82-6
Molecular Formula C12H11N
Molecular Weight (g/mol) 169.23
MDL Number MFCD00006352
InChI Key PCFUWBOSXMKGIP-UHFFFAOYSA-N
Synonym pyridine, 2-phenylmethyl, pyridine, 2-benzyl, 2-phenylmethyl pyridine, unii-ew2n4v60mq, 2-benzyl-pyridine, ew2n4v60mq, benzylpyridine, 2-benzylpyridine, pyridine, phenylmethyl, 5-20-07-00556 beilstein handbook reference
PubChem CID 7581
IUPAC Name 2-benzylpyridine
SMILES C(C1=CC=CC=C1)C1=CC=CC=N1

Specifications

Melting Point 10°C to 12°C
Density 1.054
Boiling Point 140°C to 143°C (11 mmHg)
Flash Point 125°C (257°F)
Refractive Index 1.579
Quantity 50 g
Beilstein 115875
Solubility Information Soluble in alcohol. Insoluble in water.
Formula Weight 169.23
Percent Purity ≥98%
Chemical Name or Material 2-Benzylpyridine
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 202-979-1
RTECSNumber US2620000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only