missing translation for 'onlineSavingsMsg'
Learn More

2-Benzylpyridine 98.0+%, TCI America™

$77.00 - $77.00

Chemical Identifiers

CAS 101-82-6
Molecular Formula C12H11N
Molecular Weight (g/mol) 169.23
MDL Number MFCD00006352
InChI Key PCFUWBOSXMKGIP-UHFFFAOYSA-N
Synonym pyridine, 2-phenylmethyl, pyridine, 2-benzyl, 2-phenylmethyl pyridine, unii-ew2n4v60mq, 2-benzyl-pyridine, ew2n4v60mq, benzylpyridine, 2-benzylpyridine, pyridine, phenylmethyl, 5-20-07-00556 beilstein handbook reference
PubChem CID 7581
IUPAC Name 2-benzylpyridine
SMILES C(C1=CC=CC=C1)C1=CC=CC=N1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B043625ML
View Documents
TCI America
B043625ML
25 mL
Each for $77.00
Only null left
 
Description

Description

Specifications

Chemical Identifiers

101-82-6
169.23
PCFUWBOSXMKGIP-UHFFFAOYSA-N
7581
C(C1=CC=CC=C1)C1=CC=CC=N1
C12H11N
MFCD00006352
pyridine, 2-phenylmethyl, pyridine, 2-benzyl, 2-phenylmethyl pyridine, unii-ew2n4v60mq, 2-benzyl-pyridine, ew2n4v60mq, benzylpyridine, 2-benzylpyridine, pyridine, phenylmethyl, 5-20-07-00556 beilstein handbook reference
2-benzylpyridine

Specifications

101-82-6
Yellow
C12H11N
25 mL
PCFUWBOSXMKGIP-UHFFFAOYSA-N
2-benzylpyridine
7581
≥98.0% (GC,T)
2-Benzylpyridine
10°C
277°C
MFCD00006352
pyridine, 2-phenylmethyl, pyridine, 2-benzyl, 2-phenylmethyl pyridine, unii-ew2n4v60mq, 2-benzyl-pyridine, ew2n4v60mq, benzylpyridine, 2-benzylpyridine, pyridine, phenylmethyl, 5-20-07-00556 beilstein handbook reference
C(C1=CC=CC=C1)C1=CC=CC=N1
169.23
169.23
Liquid
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (9)-1311

RTECSNumber : US2620000

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'