missing translation for 'onlineSavingsMsg'
Learn More

2-Benzylpyridine 98.0+%, TCI America™

Catalog No. B043625ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B043625ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B043625ML Supplier TCI America Supplier No. B043625ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 101-82-6
Molecular Formula C12H11N
Molecular Weight (g/mol) 169.23
MDL Number MFCD00006352
InChI Key PCFUWBOSXMKGIP-UHFFFAOYSA-N
Synonym pyridine, 2-phenylmethyl, pyridine, 2-benzyl, 2-phenylmethyl pyridine, unii-ew2n4v60mq, 2-benzyl-pyridine, ew2n4v60mq, benzylpyridine, 2-benzylpyridine, pyridine, phenylmethyl, 5-20-07-00556 beilstein handbook reference
PubChem CID 7581
IUPAC Name 2-benzylpyridine
SMILES C(C1=CC=CC=C1)C1=CC=CC=N1

Specifications

Melting Point 10°C
Color Yellow
Boiling Point 277°C
Quantity 25 mL
Formula Weight 169.23
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-Benzylpyridine
EINECSNumber (9)-1311
RTECSNumber US2620000
TSCA Yes