missing translation for 'onlineSavingsMsg'
Learn More

2-Benzyloxyphenylboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. B45901G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B45901G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B45901G Supplier TCI America Supplier No. B45901G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 190661-29-1
Molecular Formula C13H13BO3
MDL Number MFCD01632206
InChI Key MCAIDINWZOCYQK-UHFFFAOYSA-N
PubChem CID 2773253

Specifications

Melting Point 110°C
Color White
Quantity 1 g
Synonym 2-benzyloxyphenylboronic acid, 2-benzyloxy phenylboronic acid, 2-benzyloxybenzeneboronic acid, 2-benzyloxy phenyl boronic acid, 2-benzyloxyphenyl boronic acid, 2-benzyloxyphenylborornic acid, 2-benzyloxy phenyl boranediol, o-benzyloxyphenylboronic acid, boronic acid, 2-phenylmethoxy phenyl
SMILES B(C1=CC=CC=C1OCC2=CC=CC=C2)(O)O
IUPAC Name (2-phenylmethoxyphenyl)boronic acid
Molecular Weight (g/mol) 228.054
Formula Weight 228.05
Physical Form Crystalline Powder
Chemical Name or Material 2-Benzyloxyphenylboronic Acid (contains varying amounts of Anhydride)